Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%

Supplier: VWR Chemicals
Danger

Synonyms: Ethyl alcohol

Filtered through a 0.2 µm filter, packaged under nitrogen.

83672.290EA 173 EUR
83672.290 83672.230_T 83672.230 83672.269
Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
Ethanol

Bottle with a septum cap featuring six separate re-sealable puncture points


Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78.3 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

Assay Min. 99.8 %
Acidity Max. 0.0005 meq/g
Residue on evaporation Max. 0.0005 %
Water Max. 0.0030 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR