Deuterium oxide (99.90% D) for NMR spectroscopy

Supplier: VWR Chemicals

Synonyms: Water-D2, Heavy water

84541.0500EA 1450 EUR
84541.0500 84541.0011 84541.0025 84541.0100
Deuterium oxide (99.90% D) for NMR spectroscopy
Deuterium oxide
Formula: D₂O
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Specification Test Results

Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.90 %
1H-NMR Conforms to reference


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR