VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
85410.601EA 14.2 EUR
85410.601 85411.601 85412.601 85413.601 85414.601 85415.601 85416.601 85417.601 85418.601 85419.601 85420.601 85421.601 85422.601 85423.601
VWR® Dosatest, pH indicator test strips, non bleeding
Test Papers pH Indicator Papers and Test Strips
Dosatest test strips provide precise and rapid pH determination, with up to four different colour blocks for each pH value.
  • Easier to work with due to the long handle
  • Colour-fixed reactive pads
  • Brilliant colour chart
pH indicator is chemically bound onto the cellulose fibre preventing bleeding. This effectively protects the sample against contamination and enables measurements to be taken even in weak buffered or strong alkaline solutions.
Better access to different types of containers preventing fingers coming into contact with the sample.