Sulphuric acid 96 Nitrogen_analysis

Supplier: VWR Chemicals
Danger

85546.320EA 71.6 EUR
85546.320 85546.290
Sulphuric acid 96 Nitrogen_analysis
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 10.38 °C
Density: 1.84 kg/l
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
REACH: 01-2119458838-20

Order Now

Specification Test Results

Assay Min. 96 %
Colouration Max. 10 APHA
Ignition residue Max. 2 ppm
Substances reducing KMnO4 (as SO2) Max. 2 ppm
Total N (Nitrogen) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR