ESC (Easy Surface Checking) swabs
Supplier: LIOFILCHEM
85607 85601 85602 85603 85604 85605 85606 85608 85609 85610 85612 85614 85615 85621 85623
85607.EA 223 EUR
85607. 85601. 85602. 85603. 85604. 85605. 85606. 85608. 85609. 85610. 85612. 85614. 85615. 85621. 85623.
ESC (Easy Surface Checking) swabs
Monitoring Systems Hygiene Monitoring Systems
The ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
  • Pre-filled swab device
  • Rayon bud
  • Wide range of diluents
  • Leak proof closure
  • Transport and storage at room temperature
Certifications: Swabs conform to international standards (ISO 18593, ISO 17604).