Ammonium Test Strips, Dosatest
Supplier: VWR Chemicals
85702.601EA 55 EUR
85702.601
Ammonium Test Strips, Dosatest
Environmental Test Kits
Semi-quantitative test strips for the determination of ammonium.
  • Ideal for rapid and easy analysis of ammonium directly on-site
  • No maintenance or calibration is required
Colour change: Bright yellow to orange.
Delivery information: Delivered as 100 test strips in a tube with instruction leaflet.
Caution: Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.