Isohexane (mixture of isomers) ≥95% (by anhydrous basis), AnalaR NORMAPUR® ACS analytical reagent

Supplier: VWR Chemicals
Danger

Synonyms: 2-Methylpentane

85759.290EA 71.3 EUR
85759.290 85759.320
Isohexane (mixture of isomers) ≥95% (by anhydrous basis), AnalaR NORMAPUR® ACS analytical reagent
Isohexane (mixture of isomers)
Formula: C₆H₁₄
Boiling Pt: 60 °C (1013 hPa)
Melting Pt: –154 °C
Density: 0.653 g/cm³ (25 °C)
Flash Pt: −33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009406
CAS Number: 92112-69-1
EINECS: 295-570-2
UN: 1208
ADR: 3,II
REACH: 01-2119484651-34

Order Now

Specification Test Results

Assay (on anhydrous substance) min. 95.0%
Acidity max. 0.0003 meq/g
Evaporation residue max. 0.001%
Water max. 0.01%
n-Hexane max. 5%
Aromatics (as C6H6) max. 0.01%
Sulphur compounds (as S) max. 0.005%
Thiophene max. 0.0001%
Substances darkened by H2SO4 max. 10 APHA
Al (Aluminium) max. 0.5 ppm
Ba (Barium) max. 0.1 ppm
Ca (Calcium) max. 0.5 ppm
Cd (Cadmium) max. 0.05 ppm
Co (Cobalt) max. 0.02 ppm
Cr (Chromium) max. 0.02 ppm
Cu (Copper) max. 0.02 ppm
Fe (Iron) max. 0.1 ppm
Mg (Magnesium) max. 0.1 ppm
Mn (Manganese) max. 0.02 ppm
Ni (Nickel) max. 0.02 ppm
Pb (Lead) max. 0.1 ppm
Sn (Tin) max. 0.1 ppm
Zn (Zinc) max. 0.1 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR