Iodine 0.0241 M (0.0482 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Warning
85806.320EA 76.2 EUR
85806.320
Iodine 0.0241 M (0.0482 N) in aqueous solution, AVS TITRINORM volumetric solution
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
Merck Index: 13,05036
Order Now
Specification Test Results
Titer (20°C; real value 0.5 % accuracy) 0.02398 -> 0,02422 mol/l
Content KI 57g/l +/-1 g