Perchloric acid 65 - 71%, NORMATOM®, Ultrapure for trace metal analysis

Supplier: VWR Chemicals

85822.230EA 669 EUR
85822.230 85822.270 85822.290
Perchloric acid 65 - 71%, NORMATOM®, Ultrapure for trace metal analysis
Perchloric acid
Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 39
Melting Pt: -101
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 1873
ADR: 5.1 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (HClO4) 65 – 71% w/w
Ag (Silver) max. 100 ppt
Al (Aluminium) max. 100 ppt
As (Arsenic) max. 100 ppt
Ba (Barium) max. 100 ppt
Be (Beryllium) max. 100 ppt
Bi (Bismuth) max. 10 ppt
Ca (Calcium) max. 100 ppt
Cd (Cadmium) max. 10 ppt
Ce (Cerium) max. 10 ppt
Co (Cobalt) max. 100 ppt
Cs (Cesium) max. 10 ppt
Cu (Copper) max. 100 ppt
Dy (Dysprosium) max. 10 ppt
Er (Erbium) max. 10 ppt
Eu (Europium) max. 10 ppt
Fe (Iron) max. 100 ppt
Ga (Gallium) max. 10 ppt
Gd (Gadolinium) max. 10 ppt
Hf (Hafnium) Information only
Ho (Holmium) max. 10 ppt
In (Indium) max. 10 ppt
K (Potassium) max. 100 ppt
La (Lanthanum) max. 10 ppt
Li (Lithium) max. 100 ppt
Lu (Lutetium) max. 10 ppt
Mg (Magnesium) max. 100 ppt
Mn (Manganese) max. 100 ppt
Mo (Molybdenum) max. 100 ppt
Na (Sodium) max. 100 ppt
Nb (Niobium) Information only
Nd (Neodymium) max. 10 ppt
Ni (Nickel) max. 100 ppt
Pb (Lead) max. 10 ppt
Pd (Palladium) max. 10 ppt
Pr (Praseodymium) max. 10 ppt
Pt (Platinum) max. 100 ppt
Rb (Rubidium) max. 10 ppt
Rh (Rhodium) max. 10 ppt
Sb (Antimony) max. 100 ppt
Sc (Scandium) max. 100 ppt
Sm (Samarium) max. 10 ppt
Sn (Tin) max. 100 ppt
Sr (Strontium) max. 100 ppt
Ta (Tantalum) Information only
Tb (Terbium) max. 10 ppt
Te (Tellurium) max. 10 ppt
Th (Thorium) max. 10 ppt
Ti (Titanium) max. 100 ppt
Tl (Thallium) max. 100 ppt
Tm (Thulium) max. 10 ppt
U (Uranium) max. 10 ppt
V (Vanadium) max. 100 ppt
W (Tungsten) Information only
Y (Yttrium) max. 10 ppt
Yb (Ytterbium) max. 10 ppt
Zn (Zinc) max. 100 ppt
Zr (Zirconium) max. 100 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR