Formic acid 98 - 100%, NORMATOM® Trace_metal

Supplier: VWR Chemicals
Danger

Synonyms: Aminic Acid, Methanoic acid

85905.270EA 376 EUR
85905.270
Formic acid 98 - 100%, NORMATOM® Trace_metal
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Density: 1.22
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
UN: 1779
ADR: 8 (3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Coloration Max 10 APHA
Evaporation residue Max. 0.0002 %
Cl (Chloride) Max. 1 ppm
Total S (as SO₄) Max. 1 ppm
Ag (Silver) Max 0.5 ppb
Al (Aluminium) Max 5 ppb
As (Arsenic) Max 1 ppb
Au (Gold) Max 0.5 ppb
Ba (Barium) Max 5 ppb
Be (Beryllium) Max 0.5 ppb
Bi (Bismuth) Max 0.5 ppb
Ca (Calcium) Max 10 ppb
Cd (Cadmium) Max 0.5 ppb
Co (Cobalt) Max 0.5 ppb
Cr (Chromium) Max 1 ppb
Cu (Copper) Max 1 ppb
Fe (Iron) Max 5 ppb
Ga (Gallium) Max 0.5 ppb
Ge (Germanium) Max 0.5 ppb
Hg (Mercury) Max 1 ppb
In (Indium) Max 0.5 ppb
K (Potassium) Max 5 ppb
Li (Lithium) Max 0.5 ppb
Mg (Magnesium) Max 5 ppb
Mn (Manganese) Max 0.5 ppb
Mo (Molybdenum) Max 0.5 ppb
Na (Sodium) Max 15 ppb
Ni (Nickel) Max 1 ppb
Pb (Lead) Max 1 ppb
Pd (Palladium) Max 1 ppb
Pt (Platinum) Max 1 ppb
Sb (Antimony) Max 1 ppb
Sn (Tin) Max 0.5 ppb
Sr (Strontium) Max 0.5 ppb
Ti (Titanium) Max 0.5 ppb
Tl (Thallium) Max 0.5 ppb
U (Uranium) Max 0.5 ppb
V (Vanadium) Max 0.5 ppb
Zn (Zinc) Max 5 ppb
Zr (Zirconium) Max 0.5 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR