Toluene 99,5 GC_ASSAY_METHOD, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

M.O.C. Category N

8608-26EA 0 EUR
8608-26
Toluene 99,5 GC_ASSAY_METHOD, AR® ACS, Macron Fine Chemicals™
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use ≥ 99.5%
ACS - Assay (C₆H₅CH₃) (by GC) ≥ 99.5 %
Benzene (C₆H₆), For Information On ly ≤10
ACS - Color (APHA) ≤ 10
ACS - Residue after Evaporation ≤ 0.001 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Sulfur Compounds (as S) ≤ 0.003 %
ACS - Water (H₂O)(by KF, coulometric) ≤ 0.03 %
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR