Potassium pyroantimonate ≥99.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Warning

Synonyms: Potassium hexahydroxoantimonate, Potassium hexahydroxoantimonate(V), Potassium antimonate

86173.180EA 59.3 EUR
86173.180
Potassium pyroantimonate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Potassium pyroantimonate
Formula: KSbO₃·3H₂O
MW: 262.9 g/mol
MDL Number: MFCD00011415
CAS Number: 12208-13-8
UN: 1549
ADR: 6.1 III

Order Now

Specification Test Results

Appearance white powder
Assay (as KSbO₃) min. 99,0%
Loss on ignition (600 °C) max. 22%
Free alkali (as KOH) max. 0,8%
Sodium (Na) max. 0,2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR