Disodium hydrogen phosphate solution_SC 1/15 mol/l (1/15 M), AVS TITRINORM stock solution_buffer

Supplier: VWR Chemicals

Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic

86184.290EA 49.4 EUR
86184.290
Disodium hydrogen phosphate solution_SC 1/15 mol/l (1/15 M), AVS TITRINORM stock solution_buffer
di-Sodium hydrogen phosphate
Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250
MDL Number: MFCD00003496
CAS Number: 7558-79-4
Merck Index: 12,08805

Order Now

Specification Test Results

Sodium phosphate, dibasic, anhydrous 0.0664 – 0.0670 mol/l
The solution is produced by very exact volumetric or gravimetric procedures. A final analysis is not carried out.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR