VWR® AVS TITRINORM, Rubidium Standard Solution, 1000 mg/L Rb in dil. Nitric Acid, Standard for AAS
Supplier: VWR Chemicals
86702.180EA 35.7 EUR
86702.180 86702.260
VWR® AVS TITRINORM, Rubidium Standard Solution, 1000 mg/L Rb in dil. Nitric Acid, Standard for AAS
Standards Cationic Standards Rubidium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%