Lauric acid ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

87071.290EA 54.8 EUR
87071.290 87071.180 87071.460
Lauric acid ≥99%, GPR RECTAPUR®
Lauric acid
Formula: C₁₂H₂₄O₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 42…45 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400

Order Now

Specification Test Results

Assay Min. 99 %
Melting point 43 --> 45 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR