Ammonia solution 17% Reag. Ph. Eur. 1004701

Supplier: VWR Chemicals
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

87707.180EA 48.9 EUR
87707.180
Ammonia solution 17% Reag. Ph. Eur. 1004701
Ammonia solution

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: NH₃
MW: 35.05 g/mol
Density: 0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR