Dithizone solution_SC R2 Reag. Ph. Eur. 1033903

Supplier: VWR Chemicals

Synonyms: Dithizone,for analysis, 1,5-Diphenylthiocarbazone

87827.180EA 158 EUR
87827.180
Dithizone solution_SC R2 Reag. Ph. Eur. 1033903
Dithizone

Starting materials: Dithizone R / Chloroform R


Measured concentration: 2.99 +/- 0.5% ug/ml Hg


Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: C₆H₅NHNHCSN=NC₆H₅
MW: 256.33 g/mol
Boiling Pt: 406
Melting Pt: 168
MDL Number: MFCD00003025
CAS Number: 60-10-6
Merck Index: 13,03413

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR