(±)-α-Methoxyphenylacetic acid Reag. Ph. Eur. 1053601

Supplier: VWR Chemicals

Synonyms: DL-α-Methoxyphenylacetic acid, 2-Methoxy-2-phenylacetic acid, α-Methoxyphenylacetic acid

87869.030EA 588 EUR
87869.030
(±)-α-Methoxyphenylacetic acid Reag. Ph. Eur. 1053601
(±)-α-Methoxyphenylacetic acid
Formula: C₉H₁₀O₃
MDL Number: MFCD00064217
CAS Number: 7021-09-2
EINECS: 000-000-0
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR