Sodium carbonate 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1079301

Supplier: VWR Chemicals

Synonyms: Calcined soda, Carbonic acid disodium salt

87928.290EA 283 EUR
87928.290
Sodium carbonate 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1079301
Sodium carbonate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: Na₂CO₃
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR