Sodium borohydride ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

88983.22 88983.36 88983.09
88983.22EA 109 EUR
88983.22 88983.36 88983.09
Sodium borohydride ≥98%
Sodium borohydride

Purity: 98% min CAS No.: 16940-66-2 Molecular Formula: BH4Na Formula Weight: 37.83 Physical Form: -4+16 Mesh Granules Density: 1.074 Appearance: White Odour: Weak Melting Point: 400° dec. Merck Reference: 14,8592 UN No.: 1426 MDL No.: MFCD00003518

Formula: NaBH₄
Melting Pt: 400 °C (dec.)
Density: 1.074
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3,I
Merck Index: 14,8592

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR