VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 32 Elements in 5% Nitric Acid, for ICP
Supplier: VWR Chemicals
89186.180EA 857 EUR
89186.180
VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 32 Elements in 5% Nitric Acid, for ICP
Standards Cationic Standards Multi-Element Standards
32 Elements 100 mg/L: Ag, Al, Ba, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, Ge, In, K, Li, Mg, Mn, Mo, Na, Nb, Ni, P, Pb, Re, Sb, Si, Sn, Ta, Ti, V, W, Zn
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery information: Supplied with certificate of analysis.