Sodium hydroxide 8 mol/l (8 N) aqueous solution, AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Danger

Synonyms: Caustic soda

89745.290EA 48.6 EUR
89745.290
Sodium hydroxide 8 mol/l (8 N) aqueous solution, AVS TITRINORM Volumetric
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
MDL Number: MFCD00003548
CAS Number: 1310-73-2
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR