Methanol, CMOS for microelectronic, J.T.Baker®

Supplier: Avantor

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

9063-09EA 3740 EUR
9063-09
Methanol, CMOS for microelectronic, J.T.Baker®
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Flash Pt: 11
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3 II
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Microelectronic Use
Assay (CH₃OH) (by GC) ≥99.9 %
Color (APHA) ≤10
Acidity (µeq/g) ≤0.3
Alkalinity (µeq/g) ≤0.1
Residue after Evaporation ≤5 ppm
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Solubility in H₂O Passes Test
Chloride (Cl) ≤0.2 ppm
Heavy Metals (as Pb) ≤100.0 ppb
Phosphate (PO₄) ≤0.3 ppm
Sulfate (SO₄) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Trace Impurities - Arsenic (As) ≤5.0 ppb
Trace Impurities - Antimony (Sb) ≤5.0 ppb
Trace Impurities - Barium (Ba) ≤20.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤20.0 ppb
Trace Impurities - Calcium (Ca) ≤50.0 ppb
Trace Impurities - Chromium (Cr) ≤20.0 ppb
Trace Impurities - Cobalt (Co) ≤20 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤50 ppb
Trace Impurities - Germanium (Ge) ≤50.0 ppb
Trace Impurities - Gold (Au) ≤20 ppb
Trace Impurities - Iron (Fe) ≤50.0 ppb
Trace Impurities - Lead (Pb) ≤50 ppb
Trace Impurities - Lithium (Li) ≤50.0 ppb
Trace Impurities - Magnesium (Mg) ≤50.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤300.0 ppb
Trace Impurities - Nickel (Ni) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤50 ppb
Trace Impurities - Silicon (Si) ≤50 ppb
Trace Impurities - Silver (Ag) ≤20.0 ppb
Trace Impurities - Sodium (Na) ≤50.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤50 ppb
Trace Impurities - Titanium (Ti) ≤20 ppb
Trace Impurities - Vanadium (V) ≤300.0 ppb
Trace Impurities - Zinc (Zn) ≤50 ppb
Trace Impurities - Zirconium (Zr) ≤300.0 ppb
Particle Count at point of fill - 0.5 µm and greater ≤250 par/ml
Particle Count at point of fill - 1.0 µm and greater ≤25 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR