VWR® PESTINORM®, GC Multiresidue Pesticide Standard (Mix 3) - 25 components, CRM
Supplier: VWR Chemicals
91115.001EA 478 EUR
91115.001
VWR® PESTINORM®, GC Multiresidue Pesticide Standard (Mix 3) - 25 components, CRM
Standards Organic Standards
A standard mixture, typically used for food testing, containing the following components: 100 ug/ml each of Benfluralin [CAS:1861-40-1] ; Biphenyl [CAS:92-52-4] ; Chlorothalonil [CAS:1897-45-6] ; Dichlofluanid [CAS:1085-98-9] ; Dicloran [CAS:99-30-9] ; 3,4-Dichloroaniline [CAS:95-76-1] ; Dichlobenil [CAS:1194-65-6] ; Diphenylamine [CAS:122-39-4] ; Ethalfluralin [CAS:55283-68-6] ; Fluchloralin [CAS:33245-39-5] ; Isopropalin [CAS:33820-53-0] ; Nitralin [CAS:4726-14-1] ; Nitrofen [CAS:1836-75-5] ; Oxyfluorfen [CAS:42874-03-3] ; Pendimethalin [CAS:40487-42-1] ; Pentachloroaniline [CAS:527-20-8] ; Pentachlorobenzonitrile [CAS:20925-85-3] ; Quintozene [CAS:82-68-8] ; Prodiamine [CAS:29091-21-2] ; Profluralin [CAS:26399-36-0] ; 2,3,5,6-Tetrachloroaniline [CAS:3481-20-7] ; Tecnazene [CAS:117-18-0] ; cis-1,2,3,6-Tetrahydrophthalimide [CAS:1469-48-3] ; Tolylfluanid [CAS:731-27-1] ; Trifluralin [CAS:1582-09-8] in Toluene / Acetonitrile (99/1)
All mixes are supplied with detailed certificate of analysis, chromatogram and 100% shelf-life. The CoAs provide full traceability of preparation, including the dissolution of the raw materials (with their purity), the preparation of the intermediate solutions and the preparation of the final solutions.
Certifications: Certified Reference Material (CRM) acc. ISO 17025 / ISO 17034
Caution: Storage temperature: Freezer (–20 °C)