CF®DI Aminooxy
Supplier: Biotium
92177 92178 92179
92177.EA 349 EUR
92177. 92178. 92179.
CF®DI Aminooxy
Protein Linking and Labeling Systems
CF®DI aminooxy dyes react with aldehyde/ketone groups on proteins or other biomolecules to form a stable oxime bond. The CF®DI dyes (CF®488DI, CF®555DI and CF®647DI) are designed to match each other for molecular weight and charge. The molecular weights of the same protein conjugated to different CF®DI dye colors are close to each other and the pl of the labeled proteins remains the same.
  • React with carbonyl moieties of aldehyde or ketone groups on proteins and other biomolecules.
  • Size- and charge-matched fluorescent aminooxy dyes.
  • Available in 3 colors for multiplex analysis.
Biotium also offer original CF® Dye Aminooxy with a wide selection of colors. CF® dyes are next-generation fluorescent dyes with advantages in brightness, photostability, and water solubility compared to other fluorescent dyes.