VWR®, Pure Water viscosity standard

Supplier: VWR Chemicals

99566.180EA 221 EUR
99566.180
VWR®, Pure Water viscosity standard
Standards Viscometry Standards

Pure Water viscosity standard have certified data for kinematic viscosity, dynamic viscosity and density at 5, 20, 25 and 37 °C.


  • Certified in strict accordance with ASTM D2162 for 20, 25 and 37 °C
  • Density g/ml in accordance with ASTM D1480 for all temperatures
  • For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
  • Kinematic (cSt) and dynamic viscosity (mPa·s) given at all temperatures
  • Fully traceable to national standards
  • Ensures full compliance to ASTM and IP test method protocols
  • Tamper-evident security packaging


Pure Water Viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.


These standards are certified in strict accordance with ASTM D2162, the primary method for viscosity standards manufacture, for temperatures at 20, 25 and 37 °C. These products have been tested at primary level using master viscometers in accordance with ASTM D2162 to ensure the lowest uncertainty of measurement, thus providing standards that are certified with primary level accuracy.


Certifications: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.


Caution: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR