1,4-Dioxaspiro[4.5]decan-8-one ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal

A10408.06 A10408.14 A10408.22
A10408.06EA 40.7 EUR
A10408.06 A10408.14 A10408.22
1,4-Dioxaspiro[4.5]decan-8-one ≥97%
1,4-Dioxaspiro[4.5]decan-8-one

Purity: 97% CAS No.: 4746-97-8 Molecular Formula: C8H12O3 Formula Weight: 156.18 Melting Point: 72-75° Beilstein Reference: 117631

Formula: C₈H₁₂O₃
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00010214
CAS Number: 4746-97-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR