5-Nitro-2-furaldehyde ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Nitrofurfural

A10618.06 A10618.14 A10618.18
A10618.06EA 34.8 EUR
A10618.06 A10618.14 A10618.18
5-Nitro-2-furaldehyde ≥99%
5-Nitro-2-furaldehyde

Purity: 99% CAS No.: 698-63-5 Molecular Formula: C5H3NO4 Formula Weight: 141.09 Flash Point: 100°(212°F) Refractive Index: 1.5900 Melting Point: 34-38° Boiling Point: 228° Beilstein Reference: 120539 MDL No.: MFCD00003230

Formula: C₅H₃NO₄
MW: 141.08 g/mol
Boiling Pt: 228 °C
Melting Pt: 34…38 °C
Density: 1.349
Flash Pt: 33 °C (91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003230
CAS Number: 698-63-5
EINECS: 211-816-3
UN: 1325
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR