1,3-Propanediol ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: 1,3-Propylene glycol, 1,3-Dihydroxy propane

A10829.0B A10829.0I A10829.30
A10829.0BEA 186 EUR
A10829.0B A10829.0I A10829.30
1,3-Propanediol ≥99%
1,3-Propanediol

Purity: 99% CAS No.: 504-63-2 Molecular Formula: HO(CH2)3OH Formula Weight: 76.10 Flash Point: 140°(174°F) Refractive Index: 1.4400 Melting Point: -26° Boiling Point: 214° Solubility: Miscible with water, alcohol Beilstein Reference: 969155 Merck Reference: 14,9714 MDL No.: MFCD00002949

Formula: HO(CH₂)₃OH
MW: 76.1 g/mol
Boiling Pt: 214 °C
Melting Pt: –26 °C
Density: 1.053
Flash Pt: 140 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002949
CAS Number: 504-63-2
EINECS: 207-997-3
Merck Index: 12,09845

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR