(±)-1-Phenyl-1-propanol ≥98%

Supplier: Thermo Fisher Scientific
Warning

A10830.14 A10830.22
A10830.14EA 33.1 EUR
A10830.14 A10830.22
(±)-1-Phenyl-1-propanol ≥98%
(±)-1-Phenyl-1-propanol

Purity: 98+% CAS No.: 93-54-9 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 95°(203°F) Refractive Index: 1.5200 Boiling Point: 217-219° Beilstein Reference: 1906759 Merck Reference: 14,3768 MDL No.: MFCD00004564

Formula: C₆H₅CH(OH)CH₂CH₃
Boiling Pt: 217…219 °C
Density: 0.994
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004564
CAS Number: 93-54-9
EINECS: 202-256-0
Merck Index: 14,3768
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30e80b6acffb0d7728fba7e94ab32f00c90ce567&vid=f2d76406485c12e8edcd1452122e64d6ec3bd99d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR