α,2-Dibromotoluene ≥98%

Supplier: Thermo Fisher Scientific
Danger

A11285.09 A11285.18 A11285.30
A11285.09EA 56.5 EUR
A11285.09 A11285.18 A11285.30
α,2-Dibromotoluene ≥98%
α,2-Dibromotoluene

Purity: 98% CAS No.: 3433-80-5 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Flash Point: 110°(230°F) Refractive Index: 1.6190 Melting Point: 29-32° Boiling Point: 127-129°/19mm Beilstein Reference: 971015 UN No.: 1759 MDL No.: MFCD00000173

Formula: C₇H₆Br₂
Boiling Pt: 127…129 °C (19 mmHg)
Melting Pt: 29…32 °C
Density: 1.881 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000173
CAS Number: 3433-80-5
EINECS: 222-334-8
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR