Dibutylamine ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Di-n-butylamine, N-butyl-1-propanamine

A11671.0F A11671.AP
A11671.0FEA 114 EUR
A11671.0F A11671.AP
Dibutylamine ≥98%
Dibutylamine

Purity: 98+% CAS No.: 111-92-2 Molecular Formula: C8H19N Formula Weight: 129.25 Flash Point: 39°(102°F) Physical Form: Clear liquid Density: 0.767 Refractive Index: 1.4169 Appearance: Colourless Odour: Amine-like Melting Point: -62° Boiling Point: 159-160° Beilstein Reference: 506001 Merck Reference: 14,3032 UN No.: 2248 MDL No.: MFCD00009429

Formula: C₈H₁₉N
Boiling Pt: 159…160 °C
Melting Pt: –62 °C
Density: 0.767
Flash Pt: 39 °C (102 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009429
CAS Number: 111-92-2
EINECS: 203-921-8
UN: 2248
ADR: 8,II
Merck Index: 14,3032
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d7d86589224f8565e12dc8e6b30a8de923393f&vid=d2cc023db25785e54555fc105aa8436e65fd9cbc&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR