Diphenylphosphinic chloride ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Diphenylphosphinyl chloride

A11721.06 A11721.14 A11721.22
A11721.06EA 38 EUR
A11721.06 A11721.14 A11721.22
Diphenylphosphinic chloride ≥97%
Diphenylphosphinic chloride

Purity: 97+% CAS No.: 1499-21-4 Molecular Formula: C12H10ClOP Formula Weight: 236.64 Flash Point: 110°(230°F) Refractive Index: 1.6090 Melting Point: ca 20° Boiling Point: 210-212°/8mm UN No.: 3265 MDL No.: MFCD00002077

Formula: C₁₂H₁₀ClOP
Boiling Pt: 210…212 °C (8 mmHg)
Melting Pt: ∼ 20 °C
Density: 1.242
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002077
CAS Number: 1499-21-4
EINECS: 216-107-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR