N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride ≥98%

Supplier: Thermo Fisher Scientific
Warning

A12107.09 A12107.18 A12107.30
A12107.09EA 97.1 EUR
A12107.09 A12107.18 A12107.30
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride ≥98%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride

Purity: 98+% CAS No.: 637-01-4 Molecular Formula: C10H16N2.2HCl Formula Weight: 237.17 Melting Point: ca 228° dec. Beilstein Reference: 5132096

Formula: C₁₀H₁₈Cl₂N₂
Melting Pt: ∼ 228 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012482
CAS Number: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e955f6b2a1434ddba239f533c6611583219f8904&vid=13fb110b013db70c026f8eefeee7f08fa2568c0c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR