1-Dodecansulphonic acid sodium salt ≥99%

Supplier: Thermo Fisher Scientific

A12829.03 A12829.14 A12829.09
A12829.03EA 39.4 EUR
A12829.03 A12829.14 A12829.09
1-Dodecansulphonic acid sodium salt ≥99%
1-Dodecansulphonic acid sodium salt

Purity: 99% CAS No.: 2386-53-0 Molecular Formula: CH3(CH2)11SO3Na Formula Weight: 272.39 Melting Point: 300° Beilstein Reference: 3919536

Formula: C₁₂H₂₅NaO₃S
Melting Pt: >300 °C
Density: 1.218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007527
CAS Number: 2386-53-0
EINECS: 219-200-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR