Zinc sulphate heptahydrate ≥98%

Supplier: Thermo Fisher Scientific
Danger

A12915.0C A12915.0E A12915.36
A12915.0CEA 326 EUR
A12915.0C A12915.0E A12915.36
Zinc sulphate heptahydrate ≥98%
Zinc sulphate heptahydrate

Purity: 98% CAS No.: 7446-20-0 Molecular Formula: O4SZn.7H2O Formula Weight: 287.54 (161.43anhy) pH: 4.4-6 Melting Point: 100° UN No.: 3077 MDL No.: MFCD00149894

Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
Melting Pt: 100
Density: 1.97
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9 III
Merck Index: 14,10159

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR