2,3-Diaminonaphthalene 97%

Supplier: Thermo Fisher Scientific
Danger

A12993.03 A12993.06 A12993.14
A12993.03EA 94.1 EUR
A12993.03 A12993.06 A12993.14
2,3-Diaminonaphthalene 97%
2,3-Diaminonaphthalene

Purity: 97% CAS No.: 771-97-1 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 193-196° Solubility: Slightly soluble in water Beilstein Reference: 2206394 MDL No.: MFCD00004116

Formula: C₁₀H₁₀N₂
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 193…196 °C
Density: 1.096
Storage Temperature: Ambient
MDL Number: MFCD00004116
CAS Number: 771-97-1
EINECS: 212-241-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR