5-Methyl-2-furaldehyde ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Methylfurfural, 5-Methyl-2-Furaldehyde

A13264.22 A13264.14
A13264.22EA 112 EUR
A13264.22 A13264.14
5-Methyl-2-furaldehyde ≥98%
5-Methyl-2-furaldehyde

Purity: 98% CAS No.: 620-02-0 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 72°(161°F) Refractive Index: 1.5310 Boiling Point: 186-188° Beilstein Reference: 106895 MDL No.: MFCD00003232

Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 186…188 °C
Density: 1.106
Flash Pt: 72 °C (161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003232
CAS Number: 620-02-0
EINECS: 210-622-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR