Methyl-4-hydroxybenzoate ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

A14289.0B A14289.30 A14289.0I
A14289.0BEA 155 EUR
A14289.0B A14289.30 A14289.0I
Methyl-4-hydroxybenzoate ≥99%
Methyl 4-hydroxybenzoate

Purity: 99% CAS No.: 99-76-3 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless pH: 5.8 Melting Point: 125-128° Boiling Point: ca 280° dec. Beilstein Reference: 509801 Merck Reference: 14,6107 MDL No.: MFCD00002352

Formula: HOC₆H₄CO₂CH₃
Boiling Pt: ∼280 °C (dec.)
Melting Pt: 125…128 °C
Density: 1.1208 g/cm³ (137 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
EINECS: 202-785-7
Merck Index: 14,6107
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR