HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Hexamethyldisilazane

A15139.0F A15139.AE A15139.AP
A15139.0FEA 444 EUR
A15139.0F A15139.AE A15139.AP
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥98%
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
Boiling Pt: 126 °C
Melting Pt: –78 °C
Density: 0.774
Flash Pt: 8 °C (46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 3286
ADR: 3,II
REACH: 01-2119438176-38
Merck Index: 14,4689
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR