4-Methylbenzyl alcohol ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-(Hydroxymethyl)toluole, p-Tolylmethanol, (4-Methylphenyl)methanol

A15315.14 A15315.22 A15315.36
A15315.14EA 48.7 EUR
A15315.14 A15315.22 A15315.36
4-Methylbenzyl alcohol ≥99%
4-Methylbenzyl alcohol

Purity: 99% CAS No.: 589-18-4 Molecular Formula: C8H10O Formula Weight: 122.17 Melting Point: 57-61° Boiling Point: 217° MDL No.: MFCD00004664

Formula: C₈H₁₀O
Boiling Pt: 217 °C
Melting Pt: 57…61 °C
Density: 0.978
Storage Temperature: Ambient
MDL Number: MFCD00004664
CAS Number: 589-18-4
EINECS: 209-639-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR