p-Anisaldehyde ≥98%

Supplier: Thermo Fisher Scientific
Warning

A15364.30 A15364.0B A15364.18
A15364.30EA 49.1 EUR
A15364.30 A15364.0B A15364.18
p-Anisaldehyde ≥98%
p-Anisaldehyde

Purity: 98% CAS No.: 123-11-5 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 116°(241°F) Physical Form: Liquid Density: 1.121 Refractive Index: 1.5730 Sensitivity: Air and Light Sensitive Appearance: Colourless to pale yellow Odour: Odourless Melting Point: 0-2° Boiling Point: 247-249° Beilstein Reference: 471382 Merck Reference: 14,663 MDL No.: MFCD00003385

Formula: C₈H₈O₂
Boiling Pt: 247…249 °C
Melting Pt: 0…2 °C
Density: 1.121
Flash Pt: 116 °C (241 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003385
CAS Number: 123-11-5
EINECS: 204-602-6
Merck Index: 14,663

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR