L(+)-Histidine monohydrochloride monohydrate

Supplier: PanReac AppliChem

Synonyms: L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate

A1591.0100 A1591.0500 A1591.1000
A1591.0100EA 74 EUR
A1591.0100 A1591.0500 A1591.1000
L(+)-Histidine monohydrochloride monohydrate
L(+)-Histidine monohydrochloride monohydrate
Formula: C₆H₉N₃O₂·HCl·H₂O
Melting Pt: 259 °C
Density: 1.485 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151027
CAS Number: 5934-29-2
EINECS: 211-438-9
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR