Mercury(II) iodide ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Mercury diiodide, Mercuric iodide

A16130.22 A16130.36
A16130.22EA 110 EUR
A16130.22 A16130.36
Mercury(II) iodide ≥99%
Mercury (II) iodide

Purity: 99+% CAS No.: 7774-29-0 Molecular Formula: HgI2 Formula Weight: 454.40 Odour: Odourless Melting Point: 259° Boiling Point: 350° subl. Merck Reference: 14,5879 UN No.: 1638 MDL No.: MFCD00011044

Formula: HgI₂
Boiling Pt: 350 °C (subl.)
Melting Pt: 259 °C
Density: 6.28
Storage Temperature: Ambient
MDL Number: MFCD00011044
CAS Number: 7774-29-0
EINECS: 231-873-8
UN: 1638
ADR: 6.1,II
Merck Index: 14,5879

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR