Ammonium bromide ≥99%

Supplier: Thermo Fisher Scientific
Warning

A16258.0C A16258.0E A16258.36
A16258.0CEA 899 EUR
A16258.0C A16258.0E A16258.36
Ammonium bromide ≥99%
Ammonium bromide

Purity: 99% CAS No.: 12124-97-9 Molecular Formula: NH4Br Formula Weight: 97.97 Melting Point: 452° subl. Merck Reference: 14,505 MDL No.: MFCD00011419

Formula: H₄BrN
Melting Pt: 452 °C (subl.)
Density: 2.429
Storage Temperature: Ambient
MDL Number: MFCD00011419
CAS Number: 12124-97-9
EINECS: 235-183-8
Merck Index: 14,505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR