Tris-Acetate-EDTA (TAE) buffer, 50X
Supplier: PanReac AppliChem
A1691.1000
A1691.1000EA 78.3 EUR
A1691.1000
Tris-Acetate-EDTA (TAE) buffer, 50X
Buffers Electrophoresis Buffers Running Buffers
50X concentrated solution of 2 M Tris, 1 M glacial acetic acid and 0,5 M EDTA.Na₂, pH 8,5 ±0,2.