Tannic acid

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Gallotannin, Tannin

A17022.0B A17022.30 A17022.PG
A17022.0BEA 148 EUR
A17022.0B A17022.30 A17022.PG
Tannic acid
Tannic acid

CAS No.: 1401-55-4 Molecular Formula: C76H52O46 Formula Weight: 1701.23 Odour: Odourless pH: 3.5 Melting Point: ca 218° dec. Beilstein Reference: 8186396 Merck Reference: 14,9052 MDL No.: MFCD00066397

Formula: C₇₆H₅₂O₄₆
Melting Pt: ∼ 218 °C (dec.)
Density: >1 g/cm³ (20 °C)
Flash Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00066397
CAS Number: 1401-55-4
EINECS: 215-753-2
Merck Index: 14,9052

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR