Eosin B

Supplier: Thermo Fisher Scientific

A17377.14 A17377.22
A17377.14EA 44.4 EUR
A17377.14 A17377.22
Eosin B
Eosin B

CAS No.: 548-24-3 Molecular Formula: C20H6Br2N2Na2O9 Formula Weight: 624.08 MDL No.: MFCD00005041

Formula: C₂₀H₆Br₂N₂Na₂O₉
Melting Pt: > 310 °C
Storage Temperature: Ambient
MDL Number: MFCD00005041
CAS Number: 548-24-3
EINECS: 208-943-1
Merck Index: 12,03638

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR