(2-Bromophenyl)acetonitrile ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Bromobenzyl cyanide

A17548.06 A17548.14
A17548.06EA 42.6 EUR
A17548.06 A17548.14
(2-Bromophenyl)acetonitrile ≥97%
(2-Bromophenyl)acetonitrile

Purity: 97% CAS No.: 19472-74-3 Molecular Formula: C8H6BrN Formula Weight: 196.05 Flash Point: 113°(235°F) Refractive Index: 1.5696 Odour: Pungent Melting Point: 19° Boiling Point: 252° Beilstein Reference: 2043684 UN No.: 1694 MDL No.: MFCD00001896

Formula: C₈H₆BrN
Boiling Pt: 252 °C
Melting Pt: 19 °C
Density: 1.510
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001896
CAS Number: 19472-74-3
EINECS: 243-091-4
UN: 1694
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR