Oxalyl dichloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

A18012.0B A18012.18 A18012.22 A18012.30
A18012.0BEA 452 EUR
A18012.0B A18012.18 A18012.22 A18012.30
Oxalyl dichloride ≥98%
Oxalyl dichloride

Purity: 98% CAS No.: 79-37-8 Molecular Formula: C2Cl2O2 Formula Weight: 126.93 Physical Form: Liquid Density: 1.455 Refractive Index: 1.4290 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Acrid Melting Point: -12° Boiling Point: 63-64° Beilstein Reference: 1361988 Merck Reference: 14,6914 UN No.: 2927 MDL No.: MFCD00000704

Formula: C₂Cl₂O₂
Boiling Pt: 63…64 °C
Melting Pt: –12 °C
Density: 1.455
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 2927
ADR: 6.1,II
Merck Index: 14,6914

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR